Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Carbobenzoxy-β-alanine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

43481-0050 43481-0250
TS43481-0050EA 57 USD InStock
TS43481-0050 TS43481-0250
N-Carbobenzoxy-β-alanine 98%
N-Carbobenzoxy-beta-alanine


Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Melting Pt: 105.3…107.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00037292
CAS Number: 2304-94-1
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Melting point 100 to 110 °C
HPLC ≥97.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty