Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Carbobenzoxy-L-aspartic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

17205-0250 17205-1000
TS17205-0250EA 66 USD InStock
TS17205-0250 TS17205-1000
N-Carbobenzoxy-L-aspartic acid 98%
N-Carbobenzoxy-L-aspartic acid


Formula: C₁₂H₁₃NO₆
MW: 267.24 g/mol
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00002719
CAS Number: 1152-61-0
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Specific optical rotation +8° to +10° (20°C, 589 nm) (c=1, acetic acid)
Titration with NaOH 97.5 to 102.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty