Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Carbobenzoxy-L-proline 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

10837-0050 10837-0100
TS10837-0050EA 64 USD InStock
TS10837-0050 TS10837-0100
N-Carbobenzoxy-L-proline 99%
N-Carbobenzoxy-L-proline


Formula: C₁₃H₁₅NO₄
MW: 249.27 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00003170
CAS Number: 1148-11-4
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
HPLC ≥98.5 %
Specific optical rotation -59° to -61° (20°C, 589 nm) (c=2, CH3COOH)
Titration with NaOH ≥98.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty