Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O-Benzyl-N-(tert-butoxycarbonyl)-D-serine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

33467-0050 33467-0250
TS33467-0050EA 187 USD InStock
TS33467-0050 TS33467-0250
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine 98%
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine


Formula: C₁₅H₂₁NO₅
MW: 295.34 g/mol
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00038248
CAS Number: 47173-80-8
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Melting point 58 to 63 °C
HPLC ≥97.5 %
Specific optical rotation -20° to -24° (20 °C, 589 nm) (c=1, EtOH)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty