Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Omeprazole 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45852-0010 45852-0050 45852-0250
TS45852-0010EA 55 USD InStock
TS45852-0010 TS45852-0050 TS45852-0250
Omeprazole 98+%
Omeprazole


Formula: C₁₇H₁₉N₃O₃S
MW: 345.42 g/mol
Melting Pt: 158…159 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00083192
CAS Number: 73590-58-6
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (60°C, 4 h) (vacuum)
HPLC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty