Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O-Methyl-L-tyrosine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

16975-0010 16975-2500
TS16975-0010EA 102 USD InStock
TS16975-0010 TS16975-0050 TS16975-2500
O-Methyl-L-tyrosine 98%
O-Methyl-L-tyrosine


Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
Melting Pt: 259…261 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002604
CAS Number: 6230-11-1
Order Now
Specification Test Results
Appearance White to light beige Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -7° to -9° (20°C, 589 nm) (c=2, 1 N HCl)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty