Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O-tert-Butyl-L-serine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

43488-0010 43488-0050
TS43488-0010EA 107 USD InStock
TS43488-0010 TS43488-0050
O-tert-Butyl-L-serine 97%
O-tert-Butyl-L-serine


Formula: C₇H₁₅NO₃
MW: 161.20 g/mol
Melting Pt: 225.2…228 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066088
CAS Number: 18822-58-7
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤10.0 % (K.F.)
HPLC ≥96.0 %
Specific optical rotation -15.5° to -17.5° (20°C, 589 nm)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty