Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palatinose hydrate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

22595-0250 22595-1000 22595-5000
TS22595-0250EA 43 USD InStock
TS22595-0250 TS22595-1000 TS22595-5000
Palatinose hydrate 98%
Palatinose hydrate


Formula: C₁₂H₂₂O₁₁·xH₂O
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00076094
CAS Number: 343336-76-5
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤6 %
HPLC ≥97.5 % (ELSD)
Specific optical rotation +97° to +101° (20°C, 589 nm) (c=1, water)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty