Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Penta-O-acetyl-ɑ-D-glucopyranose 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

11991-0250 11991-1000
TS11991-0250EA 32 USD InStock
TS11991-0250 TS11991-1000
Penta-O-acetyl-ɑ-D-glucopyranose 98%
Penta-O-acetyl-alpha-D-glucopyranose


Formula: C₁₆H₂₂O₁₁
Melting Pt: 109…113 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00064071
CAS Number: 604-68-2
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation +96.5° to +103.5° (20°C, 589 nm) (c=1, CHCl3)
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty