Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Promethazine hydrochloride

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

25458-0050 25458-0250 25458-1000
TS25458-0050EA 46 USD InStock
TS25458-0050 TS25458-0250 TS25458-1000
Promethazine hydrochloride
Promethazine hydrochloride


Formula: C₁₇H₂₀N₂S
MW: 320.88 g/mol
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00012652
CAS Number: 58-33-3
UN: 3077
ADR: 9 III
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 4 hr)
Solubility (10% in water) Clear colorless to light yellow
Titration with HClO4 97.5 to 101.5 % (on dry substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty