Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

S-(Carboxymethyl)-L-cysteine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

22581-0010 22581-0050 22581-0250 22581-1000
TS22581-0010EA 20 USD InStock
TS22581-0010 TS22581-0050 TS22581-0250 TS22581-1000
S-(Carboxymethyl)-L-cysteine 98%
S-(Carboxymethyl)-L-cysteine


Formula: C₅H₉NO₄S
Melting Pt: 205…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00002614
CAS Number: 638-23-3
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 3 hrs)
TLC ≥97.5 %
Appearance of solution (1% soln. in 0.1 N HCl, gently warmed): clear colorless
Residue after ignition ≤0.2 %
Specific optical rotation -29° to -33° (20°C, 589 nm) (c=1, 10% NaHCO3)
Titration with HClO4 ≥97.5 %
Chloride (Cl) ≤1000 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty