Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sertraline hydrochloride

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

46219-0010 46219-0050
TS46219-0010EA 61 USD InStock
TS46219-0010 TS46219-0050
Sertraline hydrochloride
Sertraline hydrochloride


Formula: C₁₇H₁₇Cl₂N
Melting Pt: 245 °C
Storage Temperature: Freezer
MDL Number: MFCD00895772
CAS Number: 79559-97-0
Order Now
Specification Test Results
Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥97.5 %
Specific optical rotation +38° to +43° (20°C, 589 nm) (2 % in CH3OH)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty