Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triclabendazole 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

46196-0010 46196-0050
TS46196-0010EA 80 USD InStock
TS46196-0010 TS46196-0050
Triclabendazole 98%
Triclabendazole


Formula: C₁₄H₉Cl₃N₂OS
Melting Pt: 175…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00864519
CAS Number: 68786-66-3
Order Now
Specification Test Results
Appearance White to light yellow Crystallinje powder or crystals
Infrared spectrum Conforms
Melting point 175 to 180 °C
Loss on drying ≤1 % (1 g, 105 °C, 6 h)
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty