Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-5-oxopyrrolidine-2-carboxylate 50% (w/w) in water

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

23292-0250 23292-1000
TS23292-0250EA 62 USD InStock
TS23292-0250 TS23292-1000
Sodium-5-oxopyrrolidine-2-carboxylate 50% (w/w) in water
Sodium-5-oxopyrrolidine-2-carboxylate


Formula: C₅H₆NNaO₃
Boiling Pt: 109 °C
Density: 1.27 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066984
CAS Number: 54571-67-4
Order Now
Specification Test Results
Appearance Colorless to light yellow Solution
Water 47.5 to 52.5 % (K.F.)
Heavy metals ≤20 ppm
pH 6.8 to 7.4
Titration with HClO4 47.5 to 52.5 %
Arsenic (As) ≤2 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty