Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imipenem monohydrate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

45532-0010 45532-2500
TS45532-0010EA 881 USD InStock
TS45532-0010 TS45532-2500
Imipenem monohydrate 98%
Imipenem monohydrate


Formula: C₁₂H₁₇N₃O₄S
Melting Pt: 193…198 °C
Storage Temperature: Freezer
MDL Number: MFCD09753321
CAS Number: 74431-23-5
Order Now
Specification Test Results
Appearance Off-white to light yellow Powder
Infrared spectrum Conforms
Water 5 to 8 %
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty