Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Oxytetracycline dihydrate 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12384-0100 12384-1000
TS12384-0100EA 68 USD InStock
TS12384-0100 TS12384-1000
Oxytetracycline dihydrate 95%
Oxytetracycline dihydrate


Formula: C₂₂H₂₄N₂O₉·2H₂O
MW: 496.47 g/mol
Melting Pt: 181…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00151234
CAS Number: 6153-64-6
Order Now
Specification Test Results
Appearance Yellow to beige Crystalline powder
Infrared spectrum Conforms
Water 6 to 9 % (K.F.)
HPLC 94 to 102 % (on anhydrous substance)
Specific optical rotation on anhydrous substance

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty