Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Staurosporine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

32853-0010 32853-2500
TS32853-0010EA 990 USD InStock
TS32853-0010 TS32853-2500
Staurosporine 98%
Staurosporine


Formula: C₂₈H₂₆N₄O₃
MW: 466.541 g/mol
Melting Pt: 270 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18252446
CAS Number: 62996-74-1
Order Now
Specification Test Results
Appearance White to light yellow to light beige Powder
HPLC ≥97.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty