Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Inosine 5'-monophosphate disodium salt hydrate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

22626-0050 22626-0250 22626-1000
TS22626-0050EA 37 USD InStock
TS22626-0050 TS22626-0250 TS22626-1000
Inosine 5'-monophosphate disodium salt hydrate 97%
Inosine 5'-monophosphate disodium salt hydrate


Formula: C₁₀H₁₁N₄Na₂O₈P·xH₂O
MW: 392.17 g/mol (anhy)
Melting Pt: 175 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00150372
CAS Number: 352195-40-5
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤28.5 % (K.F.)
UV spectrophotometry molar absorptivity 12300 +/- 4%, on dry basis
Heavy metals (as Pb) ≤20 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty