Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Salbutamol sulfate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45646-0010 45646-0050
TS45646-0010EA 94 USD InStock
TS45646-0010 TS45646-0050
Salbutamol sulfate 98%
Salbutamol sulphate


Formula: (C₁₃H₂₁NO₃)₂·H₂SO₄
MW: 576.702 g/mol
Melting Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00055200
CAS Number: 51022-70-9
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty