Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Efaproxiral sodium 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

46010-0010 46010-2500
TS46010-0010EA 397 USD InStock
TS46010-0010 TS46010-2500
Efaproxiral sodium 98%
Efaproxiral sodium


Formula: C₂₀H₂₂NNaO₄
Storage Temperature: Ambient
MDL Number: MFCD02093864
CAS Number: 170787-99-2
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤13 %
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty