Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nafcillin sodium salt

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

46138-0010 46138-0050
TS46138-0010EA 113 USD InStock
TS46138-0010 TS46138-0050
Nafcillin sodium salt
Nafcillin sodium salt


Formula: C₂₁H₂₁N₂NaO₅S
MW: 436.46 g/mol
Melting Pt: >181 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00056863
CAS Number: 985-16-0
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤6 %
Titration with HClO4 ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty