Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silibinin

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

46303-0010 46303-0050
TS46303-0010EA 38 USD InStock
TS46303-0010 TS46303-0050
Silibinin
Silibinin


Formula: C₂₅H₂₂O₁₀
Melting Pt: 164…174 °C
Storage Temperature: Freezer
MDL Number: MFCD03225424
CAS Number: 22888-70-6
Order Now
Specification Test Results
Appearance Almost white to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤4 %
HPLC ≥97.5 % (mixture of A and B)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty