Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pravastatin sodium salt hydrate

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

46488-0010 46488-2500
TS46488-0010EA 450 USD InStock
TS46488-0010 TS46488-2500
Pravastatin sodium salt hydrate
Pravastatin sodium salt hydrate


Formula: C₂₃H₃₅O₇Na·xH₂O
MW: 446.51 g/mol
Melting Pt: 171…173 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00887601
CAS Number: 2244386-63-6
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤4 %
Specific optical rotation +150° to +159° (20°C, 589 nm) (0.5% in water)
Titration with HClO4 97.0 to 102.0 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty