Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-N-Boc-allylglycine 95%, 98% ee

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36284-0010 36284-5000
TS36284-0010EA 128 USD InStock
TS36284-0010 TS36284-5000
(R)-N-Boc-allylglycine 95%, 98% ee
(R)-N-Boc-allylglycine


Formula: C₁₀H₁₆NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD01320882
CAS Number: 170899-08-8
Order Now
Specification Test Results
Appearance Clear light yellow Viscous liquid or paste
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 %
Specific optical rotation -9.5° to -12.5° (20°C, 589 nm) (c=1, methanol)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty