Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cytidine-5'-monophosphate hydrate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

22621-0010 22621-0050
TS22621-0010EA 78 USD InStock
TS22621-0010 TS22621-0050
Cytidine-5'-monophosphate hydrate 99%
Cytidine-5'-monophosphate hydrate


Formula: C₉H₁₄N₃O₈P·xH₂O
MW: 323.20 g/mol (anhy)
Melting Pt: 222 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00149426
CAS Number: 63-37-6
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤7 % (105°C, 5 hrs)
HPLC ≥98.5 %
UV spectrophotometry molar absorptivity 13000 +/- 2%, on dry basis
Ratio of Absorbances 2.03 to 2.17 (A280/A260)
Heavy metals (as Pb) ≤10 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty