Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glycylglycine (Gly-Gly)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

12014-0010 12014-0250 12014-1000
TS12014-0010EA 896 USD InStock
TS12014-0010 TS12014-0250 TS12014-1000
Glycylglycine (Gly-Gly)
Proteins and Peptides
Glycylglycine (Gly-Gly) contains ≤0.1% sulphated ash, ≤100 ppm chloride and ≤5 ppm heavy metals (as Pb).

  • Infrared spectrum: Conforms
  • Loss on drying: ≤0.5% (105 °C, 4 h)
  • Titration with HClO₄: 99.0 to 101.0% (on dried basis)
  • EINECS: 209-127-8
  • Melting point: 262 to 264 °C (dec.)


Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty