Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Epitetracycline Hydrochloride, can be used as a Secondary Standard, Thermo Scientific

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

23312-1000 23312-5000
TS23312-1000EA 194 USD InStock
TS23312-1000 TS23312-5000
4-Epitetracycline Hydrochloride, can be used as a Secondary Standard, Thermo Scientific
Standards Organic Standards
CAS No.: 23313-80-6

MDL No.: MFCD00865028
GHS Symbols: GHS07, GHS08,
Melting Point: 217 °C (D)
Appearance: Yellow Crystalline powder
Storage Temperature: Freezer
Signal word: Warning


Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty