Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexyl acetate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

14850-0010 14850-0025 14850-0250
TS14850-0010EA 114 USD InStock
TS14850-0010 TS14850-0025 TS14850-0250
Hexyl acetate 99%
Hexyl acetate


Formula: C₈H₁₆O₂
Boiling Pt: 168…170 °C (760 mmHg)
Melting Pt: –80 °C
Density: 0.871 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00009524
CAS Number: 142-92-7
EINECS: 205-572-7
UN: 3272
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4110 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (25/25°C) 0.869 to 0.873
Acid value ≤1 mg KOH/g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty