Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Dimethyl-1-butene 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15272-0500 15272-2500
TS15272-0500EA 37 USD InStock
TS15272-0500 TS15272-2500
3,3-Dimethyl-1-butene 95%
3,3-Dimethyl-1-butene


Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 41 °C (760 mmHg)
Melting Pt: –115 °C
Density: 0.65 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008853
CAS Number: 558-37-2
EINECS: 209-195-9
UN: 3295
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3750 to 1.3780 (20°C, 589 nm)
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty