Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecylbenzene 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

15102-0100 15102-0500 15102-2500
TS15102-0100EA 54 USD InStock
TS15102-0100 TS15102-0500 TS15102-2500
Dodecylbenzene 97%
Dodecylbenzene


Formula: C₁₈H₃₀
Boiling Pt: 288…300 °C (760 mmHg)
Melting Pt: –7 °C
Density: 0.85 g/cm³
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00008974
CAS Number: 123-01-3
EINECS: 204-591-8
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4810 to 1.4830 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty