Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Vinylnaphthalene 97% stabilized

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

14090-0050 14090-0250
TS14090-0050EA 137 USD InStock
TS14090-0050 TS14090-0250
2-Vinylnaphthalene 97% stabilized
2-Vinylnaphthalene


Formula: C₁₂H₁₀
Boiling Pt: 135 °C (18 mmHg)
Melting Pt: 63…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004125
CAS Number: 827-54-3
EINECS: 212-573-6
Order Now
Specification Test Results
Appearance White to yellow to beige Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
Melting point 63 to 68 °C
GC ≥96.0 %
Stabilizer ≤0.5 % p-tert-Butylcatechol

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty