Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Indolinone 97%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12967-0050 12967-0250 12967-1000
TS12967-0050EA 53 USD InStock
TS12967-0050 TS12967-0250 TS12967-1000
2-Indolinone 97%, pure
2-Indolinone


Formula: C₈H₇NO
Boiling Pt: 227 °C (73 mmHg)
Melting Pt: 123…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00005711
CAS Number: 59-48-3
EINECS: 200-429-5
Order Now
Specification Test Results
Appearance Beige to yellow to orange to pink-brown Powder
Infrared spectrum Conforms
Melting point 123 to 128 °C
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty