Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Naphthylacetic acid 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

34964-0250 34964-1000 34964-5000
TS34964-0250EA 30 USD InStock
TS34964-0250 TS34964-1000 TS34964-5000
1-Naphthylacetic acid 95%
1-Naphthylacetic acid


Formula: C₁₂H₁₀O₂
Melting Pt: 126…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00004046
CAS Number: 86-87-3
EINECS: 201-705-8
Merck Index: 12,06458
Order Now
Specification Test Results
Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 126 to 133 °C
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty