Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-phenylanthranilic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

16300-0100 16300-1000 16300-5000
TS16300-0100EA 27 USD InStock
TS16300-0100 TS16300-1000 TS16300-5000
N-phenylanthranilic acid 98%
N-phenylanthranilic acid


Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
Boiling Pt: 375
Melting Pt: 182…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00002421
CAS Number: 91-40-7
EINECS: 202-066-8
Merck Index: 12,07427
Order Now
Specification Test Results
Appearance Yellow-green to grey-green Powder
Infrared spectrum Conforms
Melting point 182 to 187 °C
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty