Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iodoacetic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

12228-0250 12228-0500 12228-1000
TS12228-0250EA 93 USD InStock
TS12228-0250 TS12228-0500 TS12228-1000
Iodoacetic acid 97%
Iodoacetic acid


Formula: C₂H₃IO₂
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 79…83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002685
CAS Number: 64-69-7
EINECS: 200-590-1
UN: 2923
ADR: 8,I
Merck Index: 13,05047
Order Now
Specification Test Results
Appearance Yellow to orange-brown Powder or crystals or flakes
Infrared spectrum Conforms
Melting point 76 to 83 °C
Titration with NaOH ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty