Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethanesulfonic acid 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

36604-0050 36604-0250 36604-1000
TS36604-0050EA 32 USD InStock
TS36604-0050 TS36604-0250 TS36604-1000
Ethanesulfonic acid 95%
Ethanesulphonic acid


Formula: C₂H₆O₃S
Boiling Pt: 136…140 °C (3hPa)
Melting Pt: –17 °C
Density: 1.343 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007529
CAS Number: 594-45-6
EINECS: 209-843-0
UN: 2586
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4325 to 1.4355 (20°C, 589 nm)
Titration with NaOH ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty