Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxyphenylboronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

34466-0010 34466-0050
TS34466-0010EA 31 USD InStock
TS34466-0010 TS34466-0050
2-Methoxyphenylboronic acid 97%
2-Methoxyphenylboronic acid


Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 165
Storage Temperature: Refrigerator
MDL Number: MFCD00236047
CAS Number: 5720-06-9
Order Now
Specification Test Results
Appearance Off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 106 to 111 °C
Titration with NaOH ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty