Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Bromopropionic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

10735-0010 10735-2500
TS10735-0010EA 212 USD InStock
TS10735-0010 TS10735-2500
(±)-2-Bromopropionic acid 99%
(±)-2-Bromopropionic acid


Formula: H₃CBrCHCOOH
MW: 152.98 g/mol
Boiling Pt: 203
Melting Pt: 25
Density: 1.65 g/cm³
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004211
CAS Number: 598-72-1
EINECS: 209-947-6
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4760 (20°C, 589 nm)
HPLC ≥98.5 %
Stabilizer 90 to 150 ppm MEHQ

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty