Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyclopentylpropionic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11161-0050 11161-0250 11161-1000
TS11161-0050EA 23 USD InStock
TS11161-0050 TS11161-0250 TS11161-1000
3-Cyclopentylpropionic acid 98%
3-Cyclopentylpropionic acid


Formula: C₈H₁₄O₂
MW: 142.2
Boiling Pt: 130
Density: 0.99 g/cm³
Flash Pt: 46 °C
MDL Number: MFCD00001392
CAS Number: 140-77-2
EINECS: 205-433-0
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4580 (20°C, 589 nm)
Titration with NaOH ≥97.5 %
Absorbance ≤1.20 (1g/100ml cyclohexane) measured at the max. between 264nm and 276nm
Composition ≤0.5 % (delta 7-Stigmasterol)
Acid value ≤0.3 mg KOH/g
Peroxide value ≤10.0 meq O2/kg
Unsaponifiable matter ≤1.5 %
Sesame oil Complies

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty