Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Propylvaleric acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

20833-0250 20833-1000
TS20833-0250EA 68 USD InStock
TS20833-0250 TS20833-1000
2-Propylvaleric acid 99%
2-Propylvaleric acid


Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 220
Density: 0.92 g/cm³
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00002672
CAS Number: 99-66-1
EINECS: 202-777-3
UN: 3265
ADR: 8 III
Merck Index: 12,10049
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4260 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty