Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Phenylpropionic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

15910-0050 15910-1000 15910-5000
TS15910-0050EA 28 USD InStock
TS15910-0050 TS15910-1000 TS15910-5000
3-Phenylpropionic acid 99%
3-Phenylpropionic acid


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 280
Melting Pt: 47…50 °C
Density: 1.07 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00002771
CAS Number: 501-52-0
EINECS: 207-924-5
Merck Index: 13,04805
Order Now
Specification Test Results
Appearance White to yellow to light beige Crystalline powder or crystals or mass and/or chunks
Infrared spectrum Conforms
Melting point 46 to 49 °C
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty