Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diphenylacetic acid 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11715-0050 11715-1000 11715-5000
TS11715-0050EA 21 USD InStock
TS11715-0050 TS11715-1000 TS11715-5000
Diphenylacetic acid 99+%
Diphenylacetic acid


Formula: C₁₄H₁₂O₂
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00004251
CAS Number: 117-34-0
EINECS: 204-185-0
Merck Index: 13,03348
Order Now
Specification Test Results
Appearance White to off-white to cream Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 146 to 149 °C
Titration with NaOH ≥99.0 %
Calcium (Ca) ≤0.01 %
Copper (Cu) ≤0.003 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.02 %
Zinc (Zn) ≤0.003 %
Ferrous iron (Fe 2+) (Limit about 0.001%) Passes test
Insoluble matter ≤0.01 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty