Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl fumarate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11438-0010 11438-0100 11438-2500
TS11438-0010EA 228 USD InStock
TS11438-0010 TS11438-0100 TS11438-2500
Diethyl fumarate 98%
Diethyl fumarate


Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 219
Melting Pt: 1…2 °C
Density: 1.05 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00064455
CAS Number: 623-91-6
EINECS: 210-819-7
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4396 to 1.4416 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty