Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tannic acid ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

41999-0025 41999-5000
TS41999-0025EA 410 USD InStock
TS41999-0025 TS41999-5000
Tannic acid ACS
Tannic acid


Formula: C₇₆H₅₂O₄₆
MW: 1701.2 g/mol
Melting Pt: 218 °C (dec.)
Flash Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00066397
CAS Number: 1401-55-4
EINECS: 215-753-2
Merck Index: 12,09221
Order Now
Specification Test Results
Appearance Light brown Powder
Infrared spectrum Conforms
Loss on drying ≤12.0 % (105°C)
Identification Passes test
Residue after ignition ≤0.5 %
Zinc (Zn) ≤50 ppm
Heavy metals (as Pb) ≤30 ppm
Sugars, dextrin Passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty