Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,3S)-(+)-2,3-Butanediol 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

24773-0025 24773-5000
TS24773-0025EA 286 USD InStock
TS24773-0025 TS24773-5000
(2S,3S)-(+)-2,3-Butanediol 99%
(2S,3S)-(+)-2,3-Butanediol


Formula: C₄H₁₀O₂
Boiling Pt: 179…182 °C (760 mmHg)
Density: 0.98 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00063648
CAS Number: 19132-06-0
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4340 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation +12° to +14° (20°C, 589 nm) (neat)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty