Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl bromodifluoroacetate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

30580-0050 30580-0250
TS30580-0010EA 28 USD InStock
TS30580-0010 TS30580-0050 TS30580-0250
Ethyl bromodifluoroacetate 98%
Ethyl bromodifluoroacetate


Formula: BrF₂CCOOCH₂CH₃
MW: 202.98 g/mol
Boiling Pt: 112
Density: 1.583 g/cm³
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00042069
CAS Number: 667-27-6
EINECS: 211-567-0
UN: 2924
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3860 to 1.3890 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty