Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Adipohydrazide 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20331-0250 20331-1000 20331-5000
TS20331-0250EA 75 USD InStock
TS20331-0250 TS20331-1000 TS20331-5000
Adipohydrazide 98%
Adipohydrazide


Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Boiling Pt: 446
Melting Pt: 178…182 °C
Storage Temperature: Freezer
MDL Number: MFCD00007614
CAS Number: 1071-93-8
EINECS: 213-999-5
UN: 3077
ADR: 9 III
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 178 to 182 °C
Titration ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty