Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl fluoroacetate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

17266-0050 17266-0500
TS17266-0050EA 29 USD InStock
TS17266-0050 TS17266-0500
Ethyl fluoroacetate 97%
Ethyl fluoroacetate


Formula: C₄H₇FO₂
Boiling Pt: 119.2 °C (753 mmHg)
Density: 1.09 g/cm³
Flash Pt: 30 °C
MDL Number: MFCD00000450
CAS Number: 459-72-3
EINECS: 207-297-8
UN: 2929
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3745 to 1.3765 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty