Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl toluene-4-sulphonate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

17010-0025 17010-1000 17010-5000
TS17010-0025EA 375 USD InStock
TS17010-0025 TS17010-1000 TS17010-5000
Methyl toluene-4-sulphonate 97%
Methyl toluene-4-sulphonate


Formula: C₈H₁₀O₃S
MW: 186.23 g/mol
Boiling Pt: 292
Melting Pt: 24…30 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008417
CAS Number: 80-48-8
EINECS: 201-283-5
UN: 3261
ADR: 8 III
Order Now
Specification Test Results
Appearance White to yellow Low melting solid or liquid
Infrared spectrum Conforms
Refractive index 1.5150 to 1.5190 (20°C, 589 nm)
Melting point 24 to 30°C
Water ≤0.5 % (K.F.)
GC ≥96.0 %
Acidity ≤0.2 %
Titration excess NaOH 96.0 to 104.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty