Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoniazid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12260-0050 12260-1000 12260-5000
TS12260-0050EA 27 USD InStock
TS12260-0050 TS12260-1000 TS12260-5000
Isoniazid 99%
Isoniazid


Formula: C₆H₇N₃O
MW: 137.14 g/mol
Melting Pt: 171…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00006426
CAS Number: 54-85-3
EINECS: 200-214-6
Merck Index: 13,05205
Order Now
Specification Test Results
Appearance Colorless to white to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 170 to 174 °C
Loss on drying ≤0.5 % (1 g, 105 °C)
Titration with HClO4 ≥98.5 % (On dried substance)
UV at 345 nm A: ≤0.022
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty